MilliporeSigma
  • Downregulation of TLX induces TET3 expression and inhibits glioblastoma stem cell self-renewal and tumorigenesis.
Downregulation of TLX induces TET3 expression and inhibits glioblastoma stem cell self-renewal and tumorigenesis.
Nature communications (2016-02-04)
Qi Cui, Su Yang, Peng Ye, E Tian, Guoqiang Sun, Jiehua Zhou, Guihua Sun, Xiaoxuan Liu, Chao Chen, Kiyohito Murai, Chunnian Zhao, Krist T Azizian, Lu Yang, Charles Warden, Xiwei Wu, Massimo D'Apuzzo, Christine Brown, Behnam Badie, Ling Peng, Arthur D Riggs, John J Rossi, Yanhong Shi
ABSTRACT
Glioblastomas have been proposed to be maintained by highly tumorigenic glioblastoma stem cells (GSCs) that are resistant to current therapy. Therefore, targeting GSCs is critical for developing effective therapies for glioblastoma. In this study, we identify the regulatory cascade of the nuclear receptor TLX and the DNA hydroxylase Ten eleven translocation 3 (TET3) as a target for human GSCs. We show that knockdown of TLX expression inhibits human GSC tumorigenicity in mice. Treatment of human GSC-grafted mice with viral vector-delivered TLX shRNA or nanovector-delivered TLX siRNA inhibits tumour development and prolongs survival. Moreover, we identify TET3 as a potent tumour suppressor downstream of TLX to regulate the growth and self-renewal in GSCs. This study identifies the TLX-TET3 axis as a potential therapeutic target for glioblastoma.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Pricing and availability is not currently available.