MilliporeSigma
  • Clinical and Imaging Outcomes up to 1 Year Following Balloon Angioplasty for Isolated Penile Artery Stenoses in Patients With Erectile Dysfunction: The PERFECT-2 Study.
Clinical and Imaging Outcomes up to 1 Year Following Balloon Angioplasty for Isolated Penile Artery Stenoses in Patients With Erectile Dysfunction: The PERFECT-2 Study.
Journal of endovascular therapy : an official journal of the International Society of Endovascular Specialists (2016-09-16)
Tzung-Dau Wang, Wen-Jeng Lee, Shao-Chi Yang, Po-Chih Lin, Huai-Ching Tai, Shih-Ping Liu, Chien-Hua Huang, Wen-Jone Chen, Ming-Fong Chen, Ju-Ton Hsieh
ABSTRACT
To assess the angiographic and clinical outcomes in patients with erectile dysfunction and isolated penile artery stenoses treated by balloon angioplasty. In this prospective study, 22 patients (mean age 61.0±7.6 years, range 50-79) with erectile dysfunction and 34 isolated penile artery stenoses (mean 74.9%±9.1%) were enrolled and underwent balloon angioplasty. The mean International Index for Erectile Function-5 (IIEF-5) score at baseline was 10.3±4.5. The mean lesion length was 11.1±9.0 mm (mean reference vessel diameter 1.7±0.4 mm). The primary endpoint was in-segment restenosis ≥50% by pelvic computed tomography angiography (CTA) at 8 months. The 1-year sustained clinical success (IIEF-5 score ≥22 or a ≥4-point change in the IIEF-5 score and no later decline by ≥4) was the secondary outcome measure. Procedural success was achieved in 31 (91%) of 34 stenotic lesions; there was 1 flow-limiting dissection and 2 arteries with >30% residual stenosis. At 8 months, 14 of 34 lesions in 13 of 22 patients had CTA-documented binary restenosis. At 1 year, sustained clinical success was achieved in 11 of 22 patients. Of the 9 patients not developing binary restenosis, 8 achieved sustained clinical success. Our findings establish the safety and efficacy of penile artery angioplasty for patients with erectile dysfunction and isolated penile artery stenoses. They also highlight the unmet need for a more enduring treatment strategy for penile artery stenotic disease.