MilliporeSigma
  • Developmental competence and expression profile of genes in buffalo (Bubalus bubalis) oocytes and embryos collected under different environmental stress.
Developmental competence and expression profile of genes in buffalo (Bubalus bubalis) oocytes and embryos collected under different environmental stress.
Cytotechnology (2016-09-22)
E M Sadeesh, P Sikka, A K Balhara, S Balhara
ABSTRACT
The study examined the effects of different environmental stress on developmental competence and the relative abundance (RA) of various gene transcripts in oocytes and embryos of buffalo. Oocytes collected during cold period (CP) and hot period (HP) were matured, fertilized and cultured in vitro to blastocyst hatching stage. The mRNA expression patterns of genes implicated in developmental competence (OCT-4, IGF-2R and GDF-9), heat shock (HSP-70.1), oxidative stress (MnSOD), metabolism (GLUT-1), pro-apoptosis (BAX) and anti-apoptosis (BCL-2) were evaluated in immature and matured oocytes as well as in pre-implantation stage embryos. Oocytes reaching MII stage, cleavage rates, blastocyst yield and hatching rates increased (P < 0.05) during the CP. In MII oocytes and 2-cell embryos, the RA of OCT-4, IGF-2R, GDF-9, MnSOD and GLUT-1 decreased (P < 0.05) during the HP. In 4-cell embryos, the RA of OCT-4, IGF-2R and BCL-2 decreased (P < 0.05) in the HP, whereas GDF-9 increased (P < 0.05). In 8-to 16-cell embryos, the RA of OCT-4 and BCL-2 decreased (P < 0. 05) in the HP, whereas HSP-70.1 and BAX expression increased (P < 0.05). In morula and blastocyst, the RA of OCT-4, IGF-2R and MnSOD decreased (P < 0.05) during the HP, whereas HSP-70.1 was increased (P < 0.05). In conclusion, deleterious seasonal effects induced at the GV-stage carry-over to subsequent embryonic developmental stages and compromise oocyte developmental competence and quality of developed blastocysts.