MilliporeSigma
  • Removal of multi-dye wastewater by the novel integrated adsorption and Fenton oxidation process in a fluidized bed reactor.
Removal of multi-dye wastewater by the novel integrated adsorption and Fenton oxidation process in a fluidized bed reactor.
Environmental science and pollution research international (2016-08-05)
Cong Lyu, Dandan Zhou, Jun Wang
ABSTRACT
Traditionally, a few processes have to be employed in sequence for multi-dye removal, due to the different physical and chemical characteristics of the dyes. In this study, we innovatively developed an integrated adsorption and Fenton oxidation fluidized bed reactor (FBR) based on the hydraulic classification theory, which could efficiently remove dispersed red, acid yellow, and reactive brilliant dyes. The fluidized solids such as ceramsite and activated carbon could be separately fluidized at the bottom and the top part of the FBR, respectively. As a result, Fenton oxidization of dyes was promoted by the fluidization of ceramsite and activated carbon. Besides, adsorption of activated carbon could synergistically act on the dyes. The results showed that the removal efficiencies of acid yellow 2G, disperse red 60, and reactive brilliant blue X-BR could reach 100, 79.8, and 84.9 % in 10 min, respectively. Lots of intermediates with unsaturated bonds were generated during Fenton reaction, which was further removed by adsorption of activated carbon. Consequently, a high COD removal of 93 % was obtained. Interestingly, some of Fe
Sigma-Aldrich
Acid Yellow 17, Dye content 60 %
Pricing and availability is not currently available.