- Synthesis, isolation, and characterization of 2'-paclitaxel glycinate: an application of the Bsmoc protecting group.
Synthesis, isolation, and characterization of 2'-paclitaxel glycinate: an application of the Bsmoc protecting group.
The Journal of organic chemistry (2003-06-07)
Richard B Greenwald, Hong Zhao, Prasanna Reddy
PMID12790596
ABSTRACT
The relevance of the Bsmoc protecting group in the synthesis of moisture- and base-sensitive compounds has been demonstrated by the preparation of 2'-paclitaxel glycinate with use of the solid anhydrous base, 4-piperidinopiperidine in DCM solvent.