MilliporeSigma
  • Transfer hydrogenation using recyclable polyurea-encapsulated palladium: efficient and chemoselective reduction of aryl ketones.
Transfer hydrogenation using recyclable polyurea-encapsulated palladium: efficient and chemoselective reduction of aryl ketones.
Chemical communications (Cambridge, England) (2003-04-22)
Jin-Quan Yu, Hai-Chen Wu, Chandrashekar Ramarao, Jonathan B Spencer, Steven V Ley
ABSTRACT
A robust and recyclable palladium catalyst [Pd0EnCat] has been prepared by ligand exchange of polyurea-encapsulated palladium(II) acetate with formic acid, resulting in deposition of Pd(0) in the support material; Pd0EnCat is shown to be a highly efficient transfer hydrogenation catalyst for chemoselective reduction of a wide range of aryl ketones to benzyl alcohols.