MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Xanthone natural products via N-heterocyclic carbene catalysis: total synthesis of atroviridin.
Xanthone natural products via N-heterocyclic carbene catalysis: total synthesis of atroviridin.
The Journal of organic chemistry (2011-04-08)
Yumiko Suzuki, Yoshinori Fukuta, Shinya Ota, Masayo Kamiya, Masayuki Sato
ABSTRACT
The total synthesis of atroviridin has been accomplished by a linear route involving the N-heterocyclic carbene (NHC)-catalyzed aroylation of the fluorobenzene derivative, Claisen cyclization of the O-propargylated benzophenones, and intramolecular 1,4-addition of the quinone intermediates. The result provides a viable route to xanthone natural products.