MilliporeSigma
  • Cardioprotective effect of des-Aspartate-angiotensin-I (DAA-I) on cytokine gene expression profile in ligation model of myocardial infarction.
Cardioprotective effect of des-Aspartate-angiotensin-I (DAA-I) on cytokine gene expression profile in ligation model of myocardial infarction.
Life sciences (2006-01-21)
Abd Jalil Rufaihah, Husnain Kh Haider, Kwoon M Sim, Ping Z Ding, LiLi Beth Ramos, Shujia Jiang, Eugene K W Sim
ABSTRACT
We investigate the influence of des-Aspartate-angiotensin-I (DAA-I) on the cytokine expression profile in a rodent model of myocardial infarction. Myocardial infarction model was created in female Wistar rats by coronary artery ligation. Animals were randomized to receive intravenously either a daily dose of 1.2 mug DAA-I/kg body weight (group 1; n = 60) or saline (group 2; n = 60) for 14 days after infarction. Heart function was assessed by echocardiography. Animals were euthanized at 1, 3, 7, 14 and 31 days. Morphometric analysis using tetrazolium chloride staining revealed that infarct size was reduced by 32.2% (p < 0.05) in group 1 after 14 days of DAA-I treatment. Left ventricular ejection fraction in group 1 improved significantly (73.4%) as compared to group 2 (47.7%; p < 0.001). Immunostaining for immune cells at the infarct site showed that CD8+ lymphocytes infiltration at the infarct site declined in group 1 (15 +/- 5 cells) as compared to group 2 (50 +/- 6 cells; p < 0.001). Infiltration of monocytes and macrophages remained high at day 14 in group 2 (126 +/- 40 cells) as compared to group 1 (49 +/- 11 cells; p = 0.006). Multiplex PCR was done for differential gene expression of various pro-inflammatory cytokines. IL-6, TNF-alpha, TGF-beta and GM-CSF expression were significantly down-regulated in the infarct, peri-infarct and contra-lateral zones of the left ventricle in group 1 as compared to group 2. IL-6, TGF-beta and GM-CSF expression started to decline from day 1 of DAA-I treatment while TNF-alpha expression only reduced after 7 days of DAA-I treatment. We conclude that DAA-I prevented infarct expansion through suppression of inflammatory cytokines and immune cell infiltration in the infarct region.