MilliporeSigma
  • Synthesis of Amino-ADT Provides Access to Hydrolytically Stable Amide-Coupled Hydrogen Sulfide Releasing Drug Targets.
Synthesis of Amino-ADT Provides Access to Hydrolytically Stable Amide-Coupled Hydrogen Sulfide Releasing Drug Targets.
Synlett : accounts and rapid communications in synthetic organic chemistry (2016-07-12)
Matthew D Hammers, Loveprit Singh, Leticia A Montoya, Alan D Moghaddam, Michael D Pluth
ABSTRACT
As additional physiological functions of hydrogen sulfide (H