MilliporeSigma
  • Defluorinated sparfloxacin as a new photoproduct identified by liquid chromatography coupled with UV detection and tandem mass spectrometry.
Defluorinated sparfloxacin as a new photoproduct identified by liquid chromatography coupled with UV detection and tandem mass spectrometry.
Antimicrobial agents and chemotherapy (1998-05-21)
M Engler, G Rüsing, F Sörgel, U Holzgrabe
ABSTRACT
Photodegradation of sparfloxacin was observed by means of high-pressure liquid chromatography with UV detection and liquid chromatography coupled with UV detection and tandem mass spectrometry (LC-MS/MS). Three products were detected. Comparison with an independently synthesized derivative of sparfloxacin revealed the structure of one product which is believed to be 8-desfluorosparfloxacin. The second product is likely to be formed by the splitting off of a fluorine and a cyclopropyl ring. Thus, photodefluorination of quinolone antibacterial agents is found and proved for the first time by LC-MS/MS.