MilliporeSigma
  • Enantioselective organocatalytic cyclopropanations. The identification of a new class of iminium catalyst based upon directed electrostatic activation.
Enantioselective organocatalytic cyclopropanations. The identification of a new class of iminium catalyst based upon directed electrostatic activation.
Journal of the American Chemical Society (2005-03-10)
Roxanne K Kunz, David W C MacMillan
ABSTRACT
A new method for enantioselective organocatalytic cyclopropanation is described. This study outlines the identification of a new class of iminium catalyst based on the concept of directed electrostatic activation (DEA). This novel organocatalytic mechanism exploits dual activation of ylide and enal substrates through a proposed electrostatic activation and stereodirected protocol. Formation of trisubstituted cyclopropanes with high levels of enantio- and diastereoinduction is accomplished for a variety of alpha,beta-unsaturated aldehydes and sulfonium ylides. In addition, mechanistic studies have found that this cyclopropanation reaction exhibits enantioselectivity and reactivity profiles that are in accord with the proposed DEA step.
Sigma-Aldrich
(S)-(−)-Indoline-2-carboxylic acid, 99%