MilliporeSigma
  • Sulfoximines as ligands in copper-catalyzed asymmetric vinylogous Mukaiyama-type aldol reactions.
Sulfoximines as ligands in copper-catalyzed asymmetric vinylogous Mukaiyama-type aldol reactions.
Organic letters (2006-03-10)
Pauline Rémy, Martin Langner, Carsten Bolm
ABSTRACT
[reaction: see text] gamma,delta-Unsaturated alpha-hydroxy diesters with quaternary centers have been obtained with up to 99% ee in high yields using catalysts prepared from copper(II) triflate and C(1)-symmetric aminosulfoximines.