MilliporeSigma
  • Crystal structure of geneticin bound to a bacterial 16S ribosomal RNA A site oligonucleotide.
Crystal structure of geneticin bound to a bacterial 16S ribosomal RNA A site oligonucleotide.
Journal of molecular biology (2003-02-19)
Quentin Vicens, Eric Westhof
ABSTRACT
Aminoglycosides are antibacterial molecules that decrease translation accuracy by binding to the decoding aminoacyl-tRNA site (A site) on 16S ribosomal RNA. We have solved the crystal structure of an RNA fragment containing the A site bound to geneticin at 2.40A resolution. Geneticin, also known as G418, is a gentamicin-related aminoglycoside: it contains three rings that are functionalized by hydroxyl, ammonium and methyl groups. The detailed comparison of the distinctive behaviour of geneticin (binding to pro- and eukaryotic A sites) with the crystallographic, biochemical and microbiological results obtained so far for aminoglycoside-A site complexes offers new insights on the system. The two sugar rings constituting the neamine part common to most of the aminoglycosides bind to the A site, as already observed in the crystal structures solved previously with paromomycin and tobramycin. The essential hydrogen bonds involving ring I (to A1408) and ring II (to the phosphate oxygen atoms of the bulged adenine bases 1492 and 1493 and to G1494) are conserved and additional contacts are observed from ring III (to phosphate oxygen atoms of G1405 and U1406). The present work illustrates a molecular basis of the range in sensitiveness exhibited by geneticin towards common point A site mutations associated to resistance phenotypes. In addition, analysis and comparisons of the structures cast light on the role played by the conserved U1406.U1495 pair in the recognition of the A site by aminoglycosides.
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Pricing and availability is not currently available.