MilliporeSigma
  • Optogenetic approaches addressing extracellular modulation of neural excitability.
Optogenetic approaches addressing extracellular modulation of neural excitability.
Scientific reports (2016-04-06)
Emily A Ferenczi, Johannes Vierock, Kyoko Atsuta-Tsunoda, Satoshi P Tsunoda, Charu Ramakrishnan, Christopher Gorini, Kimberly Thompson, Soo Yeun Lee, Andre Berndt, Chelsey Perry, Sonja Minniberger, Arend Vogt, Joanna Mattis, Rohit Prakash, Scott Delp, Karl Deisseroth, Peter Hegemann
ABSTRACT
The extracellular ionic environment in neural tissue has the capacity to influence, and be influenced by, natural bouts of neural activity. We employed optogenetic approaches to control and investigate these interactions within and between cells, and across spatial scales. We began by developing a temporally precise means to study microdomain-scale interactions between extracellular protons and acid-sensing ion channels (ASICs). By coupling single-component proton-transporting optogenetic tools to ASICs to create two-component optogenetic constructs (TCOs), we found that acidification of the local extracellular membrane surface by a light-activated proton pump recruited a slow inward ASIC current, which required molecular proximity of the two components on the membrane. To elicit more global effects of activity modulation on 'bystander' neurons not under direct control, we used densely-expressed depolarizing (ChR2) or hyperpolarizing (eArch3.0, eNpHR3.0) tools to create a slow non-synaptic membrane current in bystander neurons, which matched the current direction seen in the directly modulated neurons. Extracellular protons played contributory role but were insufficient to explain the entire bystander effect, suggesting the recruitment of other mechanisms. Together, these findings present a new approach to the engineering of multicomponent optogenetic tools to manipulate ionic microdomains, and probe the complex neuronal-extracellular space interactions that regulate neural excitability.
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid