MilliporeSigma
  • Facile preparation and functionalization of chiral stabilized ylides from common chiral auxiliaries using triphenyl- phosphoranylideneketene (the Bestmann ylide) and their use in Wittig reactions.
Facile preparation and functionalization of chiral stabilized ylides from common chiral auxiliaries using triphenyl- phosphoranylideneketene (the Bestmann ylide) and their use in Wittig reactions.
The Journal of organic chemistry (2006-11-04)
Robert K Boeckman, Xinyi Song, Joseph E Pero
ABSTRACT
Camphor-derived lactams and other related chiral controllers have been found to react with the Bestmann ylide (triphenylphosphoranylideneketene) upon heating in toluene. The resulting parent ylides provide convenient access to a structurally diverse set of chiral stabilized ylides via functionalization. The utility of these chiral ylides for Wittig reactions has been briefly investigated and the effects of alpha-substitution noted.
Sigma-Aldrich
(S)-4-Benzyl-2-oxazolidinone, 99%
Pricing and availability is not currently available.