MilliporeSigma
  • Analysis of circulating forms of proBNP and NT-proBNP in patients with severe heart failure.
Analysis of circulating forms of proBNP and NT-proBNP in patients with severe heart failure.
Clinical chemistry (2008-03-15)
Angelika Hammerer-Lercher, Bernhard Halfinger, Bettina Sarg, Johannes Mair, Bernd Puschendorf, Andrea Griesmacher, Norberto A Guzman, Herbert H Lindner
ABSTRACT
The specific forms of pro-B-type natriuretic peptide (proBNP) that occur in human blood are not yet clear. We demonstrated the presence of several proBNP forms in human plasma with a new affinity chromatography method that can be used in combination with nano-liquid chromatography electrospray ionization tandem mass spectrometry (nano-LC-ESI-MS/MS). For affinity chromatography, we coupled Fab' fragments of polyclonal sheep antibodies specific for N-terminal proBNP (NT-proBNP) epitope 1-21 to silica beads. We connected a column (10 mm x 0.8 mm inner diameter) packed with these beads to a trypsin reactor and used a preconcentrator in combination with a fritless nanospray column to perform MS analyses of proBNP forms in preextracted and non-preextracted samples of plasma from patients with severe heart failure (HF). We used Western blotting in deglycosylation experiments to confirm the shifts in proBNP and NT-proBNP masses. Tandem MS experiments demonstrated the presence of both NT-proBNP and circulating proBNP in preextracted samples of plasma from patients with severe HF, and Western blotting analyses revealed 2 bands of approximately 23 kDa and 13 kDa that shifted after deglycosylation to positions that corresponded to the locations of recombinant proBNP and synthetic NT-proBNP. We obtained clear evidence for circulating proBNP in patients with severe HF and provided the first demonstration of O-glycosylation of NT-proBNP. The higher molecular masses for NT-proBNP and proBNP observed in the Western blotting analyses than those expected from calculations can be explained by O-glycosylation of these peptides in vivo.
Product Description
Roche
Streptavidin-POD conjugate, liquid, pkg of 1 mL
Pricing and availability is not currently available.