MilliporeSigma
  • Amyloid-beta peptide binds to microtubule-associated protein 1B (MAP1B).
Amyloid-beta peptide binds to microtubule-associated protein 1B (MAP1B).
Neurochemistry international (2007-12-15)
Goar Gevorkian, Alfonso Gonzalez-Noriega, Gonzalo Acero, Jorge Ordoñez, Colette Michalak, Maria Elena Munguia, Tzipe Govezensky, David H Cribbs, Karen Manoutcharian
ABSTRACT
Extracellular and intraneuronal formation of amyloid-beta aggregates have been demonstrated to be involved in the pathogenesis of Alzheimer's disease. However, the precise mechanism of amyloid-beta neurotoxicity is not completely understood. Previous studies suggest that binding of amyloid-beta to a number of targets have deleterious effects on cellular functions. In the present study we have shown for the first time that amyloid-beta 1-42 bound to a peptide comprising the microtubule binding domain of the heavy chain of microtubule-associated protein 1B by the screening of a human brain cDNA library expressed on M13 phage. This interaction may explain, in part, the loss of neuronal cytoskeletal integrity, impairment of microtubule-dependent transport and synaptic dysfunction observed previously in Alzheimer's disease.