MilliporeSigma
  • Evidence for expression of some microtubule-associated protein 1B in neurons as a plasma membrane glycoprotein.
Evidence for expression of some microtubule-associated protein 1B in neurons as a plasma membrane glycoprotein.
Journal of neurochemistry (2000-07-19)
S L Tanner, R Franzen, H Jaffe, R H Quarles
ABSTRACT
Microtubule-associated protein (MAP) 1B is a high-molecular-weight cytoskeletal protein that is abundant in developing neuronal processes and appears to be necessary for axonal growth. Various biochemical and immunocytochemical results are reported, indicating that a significant fraction of MAP1B is expressed as an integral membrane glycoprotein in vesicles and the plasma membrane of neurons. MAP1B is present in microsomal fractions isolated from developing rat brain and fractionates across a sucrose gradient in a manner similar to synaptophysin, a well-known vesicular and plasma membrane protein. MAP1B is also in axolemma-enriched fractions (AEFs) isolated from myelinated axons of rat brain. MAP1B in AEFs and membrane fractions from cultured dorsal root ganglion neurons (DRGNs) remains membrane-associated following high-salt washes and contains sialic acid. Furthermore, MAP1B in intact DRGNs is readily degraded by extracellular trypsin and is labeled by the cell surface probe sulfosuccinimidobiotin. Immunocytochemical examination of DRGNs shows that MAP1B is concentrated in vesicle-rich varicosities along the length of axons. Myelinated peripheral nerves immunostained for MAP1B show an enrichment at the axonal plasma membrane. These observations demonstrate that some of the MAP1B in developing neurons is an integral plasma membrane glycoprotein.