MilliporeSigma
  • Straightforward synthesis of (S)- and (R)-alpha-trifluoromethyl proline from chiral oxazolidines derived from ethyl trifluoropyruvate.
Straightforward synthesis of (S)- and (R)-alpha-trifluoromethyl proline from chiral oxazolidines derived from ethyl trifluoropyruvate.
Organic letters (2006-12-15)
Grégory Chaume, Marie-Céline Van Severen, Sinisa Marinkovic, Thierry Brigaud
ABSTRACT
[Structure: see text] A concise synthesis of both enantiomers of alpha-Tfm-proline and (S)-alpha-Tfm-prolinol from ethyl trifluoropyruvate is reported. The key step is a diastereoselective allylation reaction of ethyl trifluoropyruvate and (R)-phenylglycinol-based oxazolidines or imine. The lactone obtained by cyclization of the resulting hydroxy ester proved to be a valuable intermediate for the synthesis of (S)-alpha-Tfm-allylglycine and (S)-alpha-Tfm-norvaline in enantiopure form.
Sigma-Aldrich
(R)-(−)-2-Phenylglycinol, 98%
Pricing and availability is not currently available.