MilliporeSigma
  • Hydrodynamic structure of bovine serum albumin determined by transient electric birefringence.
Hydrodynamic structure of bovine serum albumin determined by transient electric birefringence.
Biophysical journal (1975-02-01)
A K Wright, M R Thompson
ABSTRACT
Birefringence relaxation studies on bovine serum albumin (BSA) reveal transient decay described by a double exponential process. The values of the relaxation times lead to estimation of the size of the equivalent ellipsoid of revolution for BSA. Previous measurements of transient birefringence for BSA have shown a single relaxation process, since the apparatus used in obtaining those data was not fast enough to detect the faster relaxation process.
Product Description
Roche
BSA Fraction V, ≥98.5%, Ash 1%, Heavy metals < 10 ppm, stem cell culture, Mycoplasma Negative, USA origin