MilliporeSigma
Design, Synthesis and Evaluation of New Fluoroamodiaquine Analogues.
Chemical & pharmaceutical bulletin (2016-06-03)
Ana Carolina Corrêa de Sousa, Gil Mendes Viana, Nuria Cirauqui Diaz, Marianne Grilo Rezende, Filipe Fernandes de Oliveira, Raquel Pinto Nunes, Monica Farah Pereira, André Luiz Lisboa Areas, Marianos Gustavo Zalis, Valber da Silva Frutuoso, Hugo Caire de Castro Faria, Thaisa Francielle Souza Domingos, Marcelo de Pádula, Lucio Mendes Cabral, Carlos Rangel Rodrigues
ABSTRACT
Malaria is one of the most important tropical diseases; the use of amodiaquine as a current chemotherapy in the treatment of malaria has shown some problems such as hepatotoxicity and agranulocytosis. In this work we present the rational design, synthesis, and biological evaluation (antimalarial activity, cytotoxicity and genotoxicity) of four new fluoroamodiaquine analogues. The results showed significant correlation between MolDock score and IC50 values. The molecules 7b and c were the most active of the planned compounds, with lower IC50 against Plasmodium falciparum W2 strain (0.9 and 0.8 µM, respectively) and an excellent cytotoxicity profile. The present study revealed no mutagenicity or genotoxicity for the analogues. Confirming our docking results, the molecular dynamics showed that compound 7b remains stably bound to the heme group by means of π-stacking interactions between quinoline and the porphyrin ring. Based on these findings, this study may prove to be an efficient approach for the rational design of hemozoin inhibiting compounds to treat malaria.