MilliporeSigma
Whole bone marrow cell culture: A convenient protocol for the
Experimental and therapeutic medicine (2014-08-15)
Jun-Feng Liu, Zhong-Dong DU, Zhi Chen, Zhi-Xu He
ABSTRACT
The number and function of endothelial progenitor cells (EPCs) may be a predictive factor for the severity and outcome of cardiovascular disease. However, the manipulation of bone marrow mononuclear cell (BMMC) cultures for EPCs is an elaborate and difficult procedure in small experimental animals. The present study aimed to assess the feasibility of whole bone marrow cell (WBMC) culture for expanding EPCs in small experimental animals. C57BL/6 mice (age, 3-4 weeks; weight, 9.47±0.76 g) were used as the experimental animals, and WBMCs were isolated from the femora and tibiae and cultured in endothelial cell growth medium-2. A BMMC culture for EPCs was used as a control. EPC growth, phenotype and functions were assessed
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cd34