MilliporeSigma
  • Photo-induced proton gradients for the in vitro investigation of bacterial efflux pumps.
Photo-induced proton gradients for the in vitro investigation of bacterial efflux pumps.
Scientific reports (2012-03-09)
Alice Verchère, Isabelle Broutin, Martin Picard
ABSTRACT
We describe an original activity assay for membrane transport that uses the proton motive force-dependent efflux pump MexAB from Pseudomonas aeruginosa. This pump is co-reconstituted into proteoliposomes together with bacteriorhodopsin (BR), a light-activated proton pump. In this system, upon illumination with visible light, the photo-induced proton gradient created by the BR is shown to be coupled to the active transport of substrates through the pump.
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)
Pricing and availability is not currently available.