MilliporeSigma
Galectin-8 expression in laryngeal squamous cell carcinoma.
Clinical and experimental otorhinolaryngology (2009-05-13)
Geun Woo Dong, Jun Kim, Jun Hee Park, Ji Yun Choi, Sung Il Cho, Sung Chul Lim
ABSTRACT
Despite the ongoing development of treatment protocols for laryngeal squamous cell carcinoma (LSCC), the patients suffering with this malady have shown only a modestly improved outcome. This poor outcome has been attributed to the lack of therapy that's individualized to the tumor's biological properties. Various studies have showed that galectin-8 is widely expressed in tumor tissues as well as in normal tissues, and the level of the galectin-8 expression may correlate with the malignancy of human squamous cell carcinoma. The purpose of this study is to evaluate the expression of galectin-8 and to investigate its correlations with the primary stage, the nodal involvement, the clinical stage and the histologic grade of squamous cell carcinoma of the larynx. The paraffin-embedded tissue specimens from 77 patients who were diagnosed as LSCC between 1993 and 2007 were immunohistochemically stained for galectin-8. Immunohistochemical analysis showed that a strong positive expression of galectin-8 was correlated with the T-stages, the nodal stages and the clinical stages. However, the histopathologic grades were not correlated with the galectin-8 expression in LSCC. The expression of galectin-8 protein can be used as a prognostic factor for patients with LSCC.