MilliporeSigma
  • SRp20 is a proto-oncogene critical for cell proliferation and tumor induction and maintenance.
SRp20 is a proto-oncogene critical for cell proliferation and tumor induction and maintenance.
International journal of biological sciences (2010-12-24)
Rong Jia, Cuiling Li, J Philip McCoy, Chu-Xia Deng, Zhi-Ming Zheng
ABSTRACT
Tumor cells display a different profile of gene expression than their normal counterparts. Perturbations in the levels of cellular splicing factors can alter gene expression, potentially leading to tumorigenesis. We found that splicing factor SRp20 (SFRS3) is highly expressed in cancers. SRp20 regulated the expression of Forkhead box transcription factor M1 (FoxM1) and two of its transcriptional targets, PLK1 and Cdc25B, and controlled cell cycle progression and proliferation. Cancer cells with RNAi-mediated reduction of SRp20 expression exhibited G2/M arrest, growth retardation, and apoptosis. Increased SRp20 expression in rodent fibroblasts promoted immortal cell growth and transformation. More importantly, we found that SRp20 promoted tumor induction and the maintenance of tumor growth in nude mice and rendered immortal rodent fibroblasts tumorigenic. Collectively, these results suggest that increased SRp20 expression in tumor cells is a critical step for tumor initiation, progression, and maintenance.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sfrs3