MilliporeSigma
  • Protection against Mitochondrial and Metal Toxicity Depends on Functional Lipid Binding Sites in ATP13A2.
Protection against Mitochondrial and Metal Toxicity Depends on Functional Lipid Binding Sites in ATP13A2.
Parkinson's disease (2016-04-14)
Shaun Martin, Sarah van Veen, Tine Holemans, Seyma Demirsoy, Chris van den Haute, Veerle Baekelandt, Patrizia Agostinis, Jan Eggermont, Peter Vangheluwe
ABSTRACT
The late endo-/lysosomal P-type ATPase ATP13A2 (PARK9) is implicated in Parkinson's disease (PD) and Kufor-Rakeb syndrome, early-onset atypical Parkinsonism. ATP13A2 interacts at the N-terminus with the signaling lipids phosphatidic acid (PA) and phosphatidylinositol (3,5) bisphosphate (PI(3,5)P2), which modulate ATP13A2 activity under cellular stress conditions. Here, we analyzed stable human SHSY5Y cell lines overexpressing wild-type (WT) or ATP13A2 mutants in which three N-terminal lipid binding sites (LBS1-3) were mutated. We explored the regulatory role of LBS1-3 in the cellular protection by ATP13A2 against mitochondrial stress induced by rotenone and found that the LBS2-3 mutants displayed an abrogated protective effect. Moreover, in contrast to WT, the LBS2 and LBS3 mutants responded poorly to pharmacological inhibition of, respectively, PI(3,5)P2 and PA formation. We further demonstrate that PA and PI(3,5)P2 are also required for the ATP13A2-mediated protection against the toxic metals Mn(2+), Zn(2+), and Fe(3+), suggesting a general lipid-dependent activation mechanism of ATP13A2 in various PD-related stress conditions. Our results indicate that the ATP13A2-mediated protection requires binding of PI(3,5)P2 to LBS2 and PA to LBS3. Thus, targeting the N-terminal lipid binding sites of ATP13A2 might offer a therapeutic approach to reduce cellular toxicity of various PD insults including mitochondrial stress.
Sigma-Aldrich
Anti-ATP13A2 (C-terminal region) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Rotenone, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Iron(III) chloride, reagent grade, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
FIPI hydrochloride hydrate, ≥98% (HPLC), powder
Pricing and availability is not currently available.