MilliporeSigma
  • Performance of an ELISA and indirect immunofluorescence assay in serological diagnosis of zoonotic cutaneous leishmaniasis in iran.
Performance of an ELISA and indirect immunofluorescence assay in serological diagnosis of zoonotic cutaneous leishmaniasis in iran.
Interdisciplinary perspectives on infectious diseases (2014-09-02)
Bahador Sarkari, Marzieh Ashrafmansouri, GholamReza Hatam, Parvaneh Habibi, Samaneh Abdolahi Khabisi
ABSTRACT
Serological assays have been extensively evaluated for diagnosis of visceral leishmaniasis (VL) and considered as a routine method for diagnosis of VL while these methods are not properly evaluated for diagnosis of cutaneous leishmaniasis (CL). This study aimed to assess the performance of indirect immunofluorescent-antibody test (IFA) and enzyme-linked immunosorbent assay (ELISA) for serodiagnosis of cutaneous leishmaniasis in Iran. Sixty-one sera samples from parasitologically confirmed CL patients and 50 sera from healthy controls along with 50 sera from non-CL patients were collected. Antigen was prepared from promastigotes and amastigotes of Leishmania major. IFA was used to detect anti-Leishmania IgG while ELISA was used to detect anti-Leishmania IgM, total IgG, or IgG subclasses (IgG1 and 4). ELISA, for detection of total IgG and IgM, showed sensitivity of 83.6% and 84.7% and specificity of 62.7% and 54.6%, respectively. Sensitivity and specificity of ELISA for detecting IgG1 and IgG4 were 64%, 75% and 85%, 49%, respectively. Sensitivity and specificity of IFA were 91.6% and 81%. Conclusion. Findings of this study demonstrated that serological test, especially IFA, can be used for proper diagnosis of CL.
Sigma-Aldrich
Anti-Human IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution