MilliporeSigma
  • Hydrogen sulfide and resolution of acute inflammation: A comparative study utilizing a novel fluorescent probe.
Hydrogen sulfide and resolution of acute inflammation: A comparative study utilizing a novel fluorescent probe.
Scientific reports (2012-07-13)
Neil Dufton, Jane Natividad, Elena F Verdu, John L Wallace
ABSTRACT
Hydrogen sulfide is an essential gasotransmitter associated with numerous pathologies. We assert that hydrogen sulfide plays an important role in regulating macrophage function in response to subsequent inflammatory stimuli, promoting clearance of leukocyte infiltrate and reducing TNF-α levels in vivo following zymosan-challenge. We describe two distinct methods of measuring leukocyte hydrogen sulfide synthesis; methylene blue formation following zinc acetate capture and a novel fluorescent sulfidefluor probe. Comparison of these methods, using pharmacological tools, revealed they were complimentary in vitro and in vivo. We demonstrate the application of sulfidefluor probe to spectrofluorimetry, flow cytometry and whole animal imaging, to monitor the regulation of hydrogen sulfide synthesis in vivo during dynamic inflammatory processes. Both methodologies revealed that granulocyte infiltration negatively affects hydrogen sulfide synthesis. Our report offers an insight into the profile of hydrogen sulfide synthesis during inflammation and highlight opportunities raised by the development of novel fluorescent hydrogen sulfide probes.
Sigma-Aldrich
MISSION® esiRNA, targeting human CTH
Pricing and availability is not currently available.