MilliporeSigma
Galectin-8 functions as a matricellular modulator of cell adhesion.
The Journal of biological chemistry (2001-05-24)
Y Levy, R Arbel-Goren, Y R Hadari, S Eshhar, D Ronen, E Elhanany, B Geiger, Y Zick
ABSTRACT
The interaction of cells with the extracellular matrix regulates cell adhesion and motility. Here we demonstrate that different cell types adhere and spread when cultured in serum-free medium on immobilized galectin-8, a mammalian beta-galactoside-binding protein. At maximal doses, galectin-8 is equipotent to fibronectin in promoting cell adhesion and spreading. Cell adhesion to immobilized galectin-8 is mediated by sugar-protein interactions with integrins, and galectin-8 triggers integrin-mediated signaling cascades including Tyr phosphorylation of focal adhesion kinase and paxillin. Cell adhesion is potentiated in the presence of Mn(2+), whereas it is interrupted in the presence of soluble galectin-8, integrin beta(1) inhibitory antibodies, EDTA, or thiodigalactoside but not by RGD peptides. Furthermore, cells readily adhere onto immobilized monoclonal galectin-8 antibodies, which are equipotent to integrin antibodies in promoting cell adhesion. Cell adhesion to immobilized galectin-8 is partially inhibited by serum proteins, suggesting that complex formation between immobilized galectin-8 and serum components generates a matrix that is less supportive of cell adhesion. Accordingly, cell motility on immobilized galectin-8 readily takes place in the presence of serum. Truncation of the C-terminal half of galectin-8, including one of its two carbohydrate recognition domains, largely abolishes its ability to modulate cell adhesion, indicating that both carbohydrate recognition domains are required to maintain a functional form of galectin-8. Collectively, our findings implicate galectin-8 as a physiological modulator of cell adhesion. When immobilized, it functions as a matrix protein equipotent to fibronectin in promoting cell adhesion by ligation and clustering of cell surface integrin receptors. In contrast, when present in excess as a soluble ligand, galectin-8 (like fibronectin) forms a complex with integrins that negatively regulates cell adhesion. Because of its dual effects on the adhesive properties of the cells and its association with fibronectin, galectin-8 might be considered a novel type of matricellular protein.