MilliporeSigma
  • Protection against SHIV Challenge by Subcutaneous Administration of the Plant-Derived PGT121 Broadly Neutralizing Antibody in Macaques.
Protection against SHIV Challenge by Subcutaneous Administration of the Plant-Derived PGT121 Broadly Neutralizing Antibody in Macaques.
PloS one (2016-04-01)
Yvonne J Rosenberg, David C Montefiori, Celia C LaBranche, Mark G Lewis, Markus Sack, Jonathan P Lees, Xiaoming Jiang
ABSTRACT
Intravascular delivery of broadly neutralizing antibodies (bnAbs) has shown promise for prevention and treatment of HIV infection. However, multiple IV administrations in geographic locations with poor accessibility to medical care have practical limitations. We have assessed the efficacy of plant-derived PGT121 delivered subcutaneously (SC) against pre-and post-intravaginal challenge using a rigorous SHIV-SF162P3 macaque protection model. SC administered PGT121 exhibited a longer serum half-life than IV administration and was more consistent than intramuscular delivery. A dose of 3.5mg/kg PGT121 prevented infection at a minimum ID50 neutralization titer of 1:295 while 5mg/kg protected five of six macaques when delivered immediately post-challenge. These results suggest the utility of plant-derived bnAbs delivered SC for HIV prevention.
Sigma-Aldrich
Anti-Human Kappa Light Chain (Bound and Free) antibody produced in goat, affinity isolated antibody, lyophilized powder