MilliporeSigma
  • Probing the role of backbone hydrogen bonding in beta-amyloid fibrils with inhibitor peptides containing ester bonds at alternate positions.
Probing the role of backbone hydrogen bonding in beta-amyloid fibrils with inhibitor peptides containing ester bonds at alternate positions.
Biochemistry (2003-01-15)
David J Gordon, Stephen C Meredith
ABSTRACT
Protein-protein interactions are frequently mediated by stable, intermolecular beta-sheets. A number of cytokines and the HIV Protease, for example, dimerize through beta-sheet motifs. Evidence also suggests that the macromolecular assemblies of peptides and proteins in amyloid fibrils are stabilized by intermolecular beta-sheets. In this paper, we report that interfering with the backbone hydrogen bonding of an amyloidgenic peptide (Abeta16-20) by replacing amide bonds with ester bonds prevents the aggregation of the peptide. The ester bonds were incorporated in an alternating fashion so that the peptide presents two unique hydrogen bonding faces when arrayed in an extended, beta-strand conformation; one face of the peptide has normal hydrogen bonding capabilities, but the other face is missing amide protons and its ability to hydrogen bond is severely limited. Analytical ultracentrifugation experiments demonstrate that this ester peptide, Abeta16-20e, is predominantly monomeric under solution conditions, unlike the fibril-forming Abeta16-20 peptide. Abeta16-20e also inhibits the aggregation of the Abeta1-40 peptide and disassembles preformed Abeta1-40 fibrils. These results suggest that backbone hydrogen bonding is critical for the assembly of amyloid fibrils.