MilliporeSigma
  • Experimental characterization of the human non-sequence-specific nucleic acid interactome.
Experimental characterization of the human non-sequence-specific nucleic acid interactome.
Genome biology (2013-08-02)
Gerhard Dürnberger, Tilmann Bürckstümmer, Kilian Huber, Roberto Giambruno, Tobias Doerks, Evren Karayel, Thomas R Burkard, Ines Kaupe, André C Müller, Andreas Schönegger, Gerhard F Ecker, Hans Lohninger, Peer Bork, Keiryn L Bennett, Giulio Superti-Furga, Jacques Colinge
ABSTRACT
The interactions between proteins and nucleic acids have a fundamental function in many biological processes, including gene transcription, RNA homeostasis, protein translation and pathogen sensing for innate immunity. While our knowledge of the ensemble of proteins that bind individual mRNAs in mammalian cells has been greatly augmented by recent surveys, no systematic study on the non-sequence-specific engagement of native human proteins with various types of nucleic acids has been reported. We designed an experimental approach to achieve broad coverage of the non-sequence-specific RNA and DNA binding space, including methylated cytosine, and tested for interaction potential with the human proteome. We used 25 rationally designed nucleic acid probes in an affinity purification mass spectrometry and bioinformatics workflow to identify proteins from whole cell extracts of three different human cell lines. The proteins were profiled for their binding preferences to the different general types of nucleic acids. The study identified 746 high-confidence direct binders, 139 of which were novel and 237 devoid of previous experimental evidence. We could assign specific affinities for sub-types of nucleic acid probes to 219 distinct proteins and individual domains. The evolutionarily conserved protein YB-1, previously associated with cancer and drug resistance, was shown to bind methylated cytosine preferentially, potentially conferring upon YB-1 an epigenetics-related function. The dataset described here represents a rich resource of experimentally determined nucleic acid-binding proteins, and our methodology has great potential for further exploration of the interface between the protein and nucleic acid realms.
Sigma-Aldrich
Anti-Nucleolin Antibody, clone 3G4B2, culture supernatant, clone 3G4B2, Upstate®
Pricing and availability is not currently available.