MilliporeSigma
  • Mice transgenic for BAFF develop lymphocytic disorders along with autoimmune manifestations.
Mice transgenic for BAFF develop lymphocytic disorders along with autoimmune manifestations.
The Journal of experimental medicine (1999-12-10)
F Mackay, S A Woodcock, P Lawton, C Ambrose, M Baetscher, P Schneider, J Tschopp, J L Browning
ABSTRACT
The cause of many autoimmune and inflammatory diseases is unresolved, although dysregulated production of tumor necrosis factor (TNF) family members appears to be important in many cases. BAFF, a new member of the TNF family, binds to B cells and costimulates their growth in vitro. Mice transgenic for BAFF have vastly increased numbers of mature B and effector T cells, and develop autoimmune-like manifestations such as the presence of high levels of rheumatoid factors, circulating immune complexes, anti-DNA autoantibodies, and immunoglobulin deposition in the kidneys. This phenotype is reminiscent of certain human autoimmune disorders and suggests that dysregulation of BAFF expression may be a critical element in the chain of events leading to autoimmunity.