MilliporeSigma
  • Quenching-Chemiluminescence Determination of Trace Amounts of l-Tyrosine Contained in Dietary Supplement by Chemiluminescence Reaction of an Iron-Phthalocyanine Complex.
Quenching-Chemiluminescence Determination of Trace Amounts of l-Tyrosine Contained in Dietary Supplement by Chemiluminescence Reaction of an Iron-Phthalocyanine Complex.
Journal of analytical methods in chemistry (2012-05-09)
Takao Ohtomo, Shukuro Igarashi, Yoshitaka Takagai, Osamu Ohno
ABSTRACT
The chemiluminescence (CL) signal immediately appeared when a hydrogen peroxide solution was injected into an iron-phthalocyanine tetrasulfonic acid (Fe-PTS) aqueous solution. Moreover, the CL intensity of Fe-PTS decreased by adding l-tyrosine. Based on these results, the determination of trace amounts of l-tyrosine was developed using the quenching-chemiluminescence. The calibration curve of l-tyrosine was obtained in the concentration range of 2.0 × 10(-7) M to 2.0 × 10(-5) M. Moreover, the relative standard deviation (RSD) was 1.63 % (n = 5) for 2.0 × 10(-6) M l-tyrosine, and its detection limits (3σ) were 1.81 × 10(-7) M. The spike and recovery experiments for l-tyrosine were performed using a soft drink. Furthermore, the determination of l-tyrosine was applied to supplements containing various kinds of amino acids. Each satisfactory relative recovery was obtained at 98 to 102%.