MilliporeSigma
  • The proton affinity of the superbase 1,8-bis(tetramethylguanidino)naphthalene (TMGN) and some related compounds: a theoretical study.
The proton affinity of the superbase 1,8-bis(tetramethylguanidino)naphthalene (TMGN) and some related compounds: a theoretical study.
Chemistry (Weinheim an der Bergstrasse, Germany) (2002-04-05)
Borislav Kovacević, Zvonimir B Maksić
ABSTRACT
The spatial and electronic structure of the very strong neutral organic bases bis(tetramethylguanidino)naphthalene (TMGN), 4,5-bis(tetramethylguanidino)fluorene (TMGF) and some related compounds are explored by ab initio computational methods. Their affinity towards the proton is scrutinized both in the gas phase and in solution in acetonitrile. The protonation at the most basic center (the imine nitrogen) yields asymmetric and relatively strong intramolecular hydrogen bonds (IHB). It is found that the angular strain effect and steric repulsion practically vanish in TMGN which implies that its high absolute proton affinity (APA) has its origin in the inherent basicity of the guanidine fragment and a relatively strong IHB in [TMGN]H(+). The nonbonded repulsions in TMGF are higher than in TMGN, which in conjunction with a slightly stronger IHB in the corresponding conjugate acid makes it more basic: APA(TMGF)>APA(TMGN). An interesting new phenomenon is observed in both TMGN and TMGF: the proton triggers the resonance stabilization not only in the directly bonded guanidine moiety, but also in the other guanidine fragment which is more distant from the proton, albeit in a less pronounced manner. The latter feature is termed a partial protonation. This supports the hydrogen bonding and contributes to the IHB stabilization. Convincing evidence is presented that the solvent effect in acetonitrile is determined by two antagonistic factors: 1) the intrinsic (gas phase) proton affinity and 2) the size effect which is given by the ratio between the positive charge in molecular cation (conjugate acid) and the magnitude of the molecular surface. The resulting pK(a) values are given by an interplay of these factors.
Sigma-Aldrich
1,8-Bis(tetramethylguanidino)naphthalene, ≥98.0%