MilliporeSigma
  • Analysis of amino acids by gas chromatography-flame ionization detection and gas chromatography-mass spectrometry: simultaneous derivatization of functional groups by an aqueous-phase chloroformate-mediated reaction.
Analysis of amino acids by gas chromatography-flame ionization detection and gas chromatography-mass spectrometry: simultaneous derivatization of functional groups by an aqueous-phase chloroformate-mediated reaction.
Journal of chromatography. A (1994-03-04)
J Wang, Z H Huang, D A Gage, J T Watson
ABSTRACT
The one-step ethyl chloroformate derivatization of amino acids in an aqueous medium is extended with the use of a variety of alkyl chloroformate reagents. This provides a new and convenient procedure for preparing esters with different alkoxy groups. A new mechanism for esterification during chloroformate derivatization is proposed based on the formation of an intermediate mixed carboxylic-carbonic acid anhydride followed by the exchange with an alcohol. Among the different reagents investigated, isobutyl chloroformate derivatized amino acids were found to provide more sensitivity for analyses by GC-flame ionization detection and GC-MS relative to derivatives prepared by other alkyl chloroformates.
Sigma-Aldrich
Isobutyl chloroformate, 98%