MilliporeSigma
  • Determination of volatile organics in drinking water with USEPA method 524.2 and the ion trap detector.
Determination of volatile organics in drinking water with USEPA method 524.2 and the ion trap detector.
Journal of chromatographic science (1990-09-01)
J W Eichelberger, T A Bellar, J P Donnelly, W L Budde
ABSTRACT
New drinking water regulations require the monitoring of eight volatile organic compounds that have established maximum contaminant levels (MCLs) and 51 other volatile organics for which MCLs are not established. A laboratory analytical method (Method 524.2) for the determination of 58 of these compounds is investigated, and precision and accuracy data are obtained. The method uses a standard inert gas purge extraction, isolation of the volatile organics on a three-stage solid-phase trap, thermal desorption into a gas chromatograph, separation with a fused-silica capillary column, and identification and measurement with a relatively low cost, benchtop ion trap detector that functions as a mass spectrometer. At a concentration of 2 micrograms/L (2 parts per billion), the grand mean measurement accuracy for 54 compounds was 95% of the true value with a mean relative standard deviation (RSD) of 4%. At 0.2 micrograms/L (200 parts per trillion), the grand mean measurement accuracy for 52 compounds was 95% of the true value with a mean RSD of 3%.
Sigma-Aldrich
1,2-Dichlorobenzene-d4, 98 atom % D
Pricing and availability is not currently available.