MilliporeSigma
  • Subversion of autophagy in adherent invasive Escherichia coli-infected neutrophils induces inflammation and cell death.
Subversion of autophagy in adherent invasive Escherichia coli-infected neutrophils induces inflammation and cell death.
PloS one (2012-12-29)
Abderrahman Chargui, Annabelle Cesaro, Sanda Mimouna, Mohamed Fareh, Patrick Brest, Philippe Naquet, Arlette Darfeuille-Michaud, Xavier Hébuterne, Baharia Mograbi, Valérie Vouret-Craviari, Paul Hofman
ABSTRACT
Invading bacteria are recognized, captured and killed by a specialized form of autophagy, called xenophagy. Recently, defects in xenophagy in Crohn's disease (CD) have been implicated in the pathogenesis of human chronic inflammatory diseases of uncertain etiology of the gastrointestinal tract. We show here that pathogenic adherent-invasive Escherichia coli (AIEC) isolated from CD patients are able to adhere and invade neutrophils, which represent the first line of defense against bacteria. Of particular interest, AIEC infection of neutrophil-like PLB-985 cells blocked autophagy at the autolysosomal step, which allowed intracellular survival of bacteria and exacerbated interleukin-8 (IL-8) production. Interestingly, this block in autophagy correlated with the induction of autophagic cell death. Likewise, stimulation of autophagy by nutrient starvation or rapamycin treatment reduced intracellular AIEC survival and IL-8 production. Finally, treatment with an inhibitor of autophagy decreased cell death of AIEC-infected neutrophil-like PLB-985 cells. In conclusion, excessive autophagy in AIEC infection triggered cell death of neutrophils.
Product Description
Roche
Nutridoma-SP, suitable for hybridoma, solution, pkg of 100 mL (100x)