MilliporeSigma
  • Synthesis of new derivatives of 4-acylquinoxalin-2-ones with potential pharmacological activity.
Synthesis of new derivatives of 4-acylquinoxalin-2-ones with potential pharmacological activity.
Acta poloniae pharmaceutica (1994-01-01)
E Mikiciuk-Olasik, P Trzebiński, R Nowak, B Kotełko
ABSTRACT
Some of new derivatives of 4-acylquinoxalin-2-one obtained in the reaction of N,N'-bis(chloroacetyl)-4,5-dimethyl-o-phenylenediamine with primary amines are described. Compounds were tested for their anti-HIV activity and as ligands for 99mTc.
Sigma-Aldrich
4,5-Dimethyl-1,2-phenylenediamine, 98%