MilliporeSigma
  • Waterborne toxicity and diet-related effects of fungicides in the key leaf shredder Gammarus fossarum (Crustacea: Amphipoda).
Waterborne toxicity and diet-related effects of fungicides in the key leaf shredder Gammarus fossarum (Crustacea: Amphipoda).
Aquatic toxicology (Amsterdam, Netherlands) (2015-11-02)
J P Zubrod, D Englert, J Wolfram, D Wallace, N Schnetzer, P Baudy, M Konschak, R Schulz, M Bundschuh
ABSTRACT
Animals involved in leaf litter breakdown (i.e., shredders) play a central role in detritus-based stream food webs, while their fitness and functioning can be impaired by anthropogenic stressors. Particularly fungicides can affect shredders via both waterborne exposure and their diet, namely due to co-ingestion of adsorbed fungicides and shifts in the leaf-associated fungal community, on which shredders' nutrition heavily relies. To understand the relevance of these effect pathways, we used a full 2×2-factorial test design: the leaf material serving as food was microbially colonized for 12 days either in a fungicide-free control or exposed to a mixture of five current-use fungicides (sum concentration of 62.5μg/L). Similarly, the amphipod shredder Gammarus fossarum was subjected to the same treatments but for 24 days. Waterborne exposure reduced leaf consumption by ∼20%, which did not fully explain the reduction in feces production (∼30%), indicating an enhanced utilization of food to compensate for detoxification mechanisms. This may also explain the reduced feces production (∼10%) of gammarids feeding on fungicide-exposed leaves. The reduction may, however, also be caused by a decreased nutritious quality of the leaves indicated by a reduced species richness (∼40%) of leaf-associated fungi. However, compensation for these effects by Gammarus was seemingly incomplete, since both waterborne exposure and the consumption of the fungicide-affected diet drastically reduced gammarid growth (∼110% and ∼40%, respectively). Our results thus indicate that fungicide mixtures have the potential for detrimental implications in aquatic ecosystem functioning by affecting shredders via both effect pathways.
Product Description
Supelco
Quinoxyfen, PESTANAL®, analytical standard