MilliporeSigma
  • Biosynthesis of 2-acetyl-1-pyrroline in rice calli cultures: Demonstration of 1-pyrroline as a limiting substrate.
Biosynthesis of 2-acetyl-1-pyrroline in rice calli cultures: Demonstration of 1-pyrroline as a limiting substrate.
Food chemistry (2015-12-01)
Janchai Poonlaphdecha, Pascal Gantet, Isabelle Maraval, François-Xavier Sauvage, Chantal Menut, Alain Morère, Renaud Boulanger, Matthias Wüst, Ziya Gunata
ABSTRACT
The role of 1-pyrroline was studied via feeding experiments using rice calli cultures to gain further insight into the key steps of 2-acetyl-1-pyrroline (2AP) biosynthesis in rice. The origin of the acetyl donor was also studied through stable isotope labelled substrates. Incubation of fresh calli from a fragrant rice variety (Aychade) and a non-fragrant variety (Gladio×Fidji K2) with 1-pyrroline led to a significant increase in 2AP in both varieties. Importantly, the amount of 2AP in the non-fragrant variety could be greatly enhanced by this supplementation. When rice calli were fed with increasing levels of 1-pyrroline, 2AP levels increased accordingly. Our data show that 1-pyrroline is a limiting factor for 2AP synthesis in rice. Heat treatment of calli suggested that 1-pyrroline might be enzymatically acetylated. The presence of labelled 2AP in calli supplemented with [U-(13)C]glucose, sodium acetate (1,2-(13)C2) and sodium octanoate (1,2,3,4-(13)C4) suggested that these compounds are possible candidates for acetyl group-donors of 2AP, predominately in the form of intact labelled (13)C2-units.
Sigma-Aldrich
4-Aminobutyraldehyde diethyl acetal, 90%, technical grade
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Pricing and availability is not currently available.