MilliporeSigma
  • Perfluorooctane Sulfonate Plasma Half-Life Determination and Long-Term Tissue Distribution in Beef Cattle (Bos taurus).
Perfluorooctane Sulfonate Plasma Half-Life Determination and Long-Term Tissue Distribution in Beef Cattle (Bos taurus).
Journal of agricultural and food chemistry (2015-12-20)
Sara J Lupton, Kerry L Dearfield, John J Johnston, Sarah Wagner, Janice K Huwe
ABSTRACT
Perfluorooctane sulfonate (PFOS) is used in consumer products as a surfactant and is found in industrial and consumer waste, which ends up in wastewater treatment plants (WWTPs). PFOS does not breakdown during WWTP processes and accumulates in the biosolids. Common practices include application of biosolids to pastures and croplands used for feed, and as a result, animals such as beef cattle are exposed to PFOS. To determine plasma and tissue depletion kinetics in cattle, 2 steers and 4 heifers were dosed with PFOS at 0.098 mg/kg body weight and 9.1 mg/kg, respectively. Plasma depletion half-lives for steers and heifers were 120 ± 4.1 and 106 ± 23.1 days, respectively. Specific tissue depletion half-lives ranged from 36 to 385 days for intraperitoneal fat, back fat, muscle, liver, bone, and kidney. These data indicate that PFOS in beef cattle has a sufficiently long depletion half-life to permit accumulation in edible tissues.
Sigma-Aldrich
Tetrabutylammonium sulfate solution, 50 wt. % in H2O