MilliporeSigma
  • Glutathione S-transferase alpha 1 risk polymorphism and increased bilateral thalamus mean diffusivity in schizophrenia.
Glutathione S-transferase alpha 1 risk polymorphism and increased bilateral thalamus mean diffusivity in schizophrenia.
Psychiatry research (2012-09-11)
Gianfranco Spalletta, Fabrizio Piras, Paolo Gravina, Mario Lo Bello, Sergio Bernardini, Carlo Caltagirone
ABSTRACT
Oxidative damage in brain cells is one of the factors hypothesized to be involved in the pathogenesis of schizophrenia. Glutathione S-transferase (GST) A1*B polymorphism, a genotype associated with a higher risk of oxidative damage, is associated with increased frequency of schizophrenia diagnosis. Thus, here we studied Glutathione S-transferase (GST) A1 polymorphism and diffusion tensor imaging-mean diffusivity (MD) data on deep grey matter brain structures in 56 patients with Diagnostic and Statistical Manual of Mental Disorders, Fourth Edition, Text Revised (DSM-IV-TR) schizophrenia. Clinical diagnosis and psychopathological symptom severity were assessed by using the Structured Clinical Interview for DSM-IV-TR (SCID-P) and the Scales for Assessment of Positive and Negative Symptoms (SAPS and SANS). Results confirmed that patients with schizophrenia who were carriers of the GSTA1 *B risk allele had an increased MD in bilateral thalami and increased severity of auditory and global hallucinations in comparison with non-B carriers. Thus, oxidative stress associated factors may be implicated in specific mechanisms of schizophrenia such as altered microstructure of the thalami and specific psychopathological features of auditory hallucinations.
Sigma-Aldrich
GST A1-1, Recombinant Human