MilliporeSigma
  • Impact of Alternaria toxins on CYP1A1 expression in different human tumor cells and relevance for genotoxicity.
Impact of Alternaria toxins on CYP1A1 expression in different human tumor cells and relevance for genotoxicity.
Toxicology letters (2015-10-18)
G Pahlke, C Tiessen, K Domnanich, N Kahle, I A M Groh, I Schreck, C Weiss, D Marko
ABSTRACT
The Alternaria toxins alternariol (AOH) and alternariol monomethyl ether (AME) have been reported previously to act as activators of the aryl hydrocarbon receptor (AhR) in murine hepatoma cells, thus enhancing the expression of cytochrome P450 (CYP) 1A monooxygenases. Concomitantly, both benzopyrones represent substrates of CYP1A, giving rise to catecholic metabolites. The impact of AOH and AME on CYP1A expression in human cells of different tissue origin colon (HT29), esophagus (KYSE510), liver (HepG2) and their effects on cell viability, generation of reactive oxygen species (ROS) and DNA integrity were investigated. ROS production was induced by both mycotoxins in all cell lines with AOH exhibiting the highest potency in esophageal cells concomitant with the most prominent CYP1A induction level. Of note, altertoxin-II (ATX-II), the more potent DNA-damaging mutagen formed by Alternaria alternata, induces CYP1A even at significant lower concentrations. AhR-siRNA knockdown in human esophageal cells supported the hypothesis of AhR-mediated CYP1A1 induction by AOH. However, DNA damage was minor at CYP1A1-inducing AOH concentrations. AhR-depletion did not affect the DNA-damaging properties of AOH indicating no substantial impact of AhR in this regard. However, in combination with xenobiotics prone to metabolic activation by CYP1A the induction of CYP1A by Alternaria toxins deserves further attention.
Sigma-Aldrich
CypExpress 1A1 Cytochrome P450 human
Pricing and availability is not currently available.