MilliporeSigma
  • Graft-versus-leukemia antigen CML66 elicits coordinated B-cell and T-cell immunity after donor lymphocyte infusion.
Graft-versus-leukemia antigen CML66 elicits coordinated B-cell and T-cell immunity after donor lymphocyte infusion.
Clinical cancer research : an official journal of the American Association for Cancer Research (2010-05-13)
Wandi Zhang, Jaewon Choi, Wanyong Zeng, Shelby A Rogers, Edwin P Alyea, James G Rheinwald, Christine M Canning, Vladimir Brusic, Tetsuro Sasada, Ellis L Reinherz, Jerome Ritz, Robert J Soiffer, Catherine J Wu
ABSTRACT
The target antigens of graft-versus-leukemia that are tumor associated are incompletely characterized. We examined responses developing against CML66, an immunogenic antigen preferentially expressed in myeloid progenitor cells identified from a patient with chronic myelogenous leukemia who attained long-lived remission following CD4+ donor lymphocyte infusion (DLI). From this patient, CML66-reactive CD8+ T-cell clones were detected against an endogenously presented HLA-B*4403-restricted epitope (HDVDALLW). Neither CML66-specific antibody nor T-cell responses were detectable in peripheral blood before DLI. However, by 1 month after DLI, CD8+ T cells were present in peripheral blood and at 10-fold higher frequency in marrow. Subsequently, plasma antibody to CML66 developed in association with disease remission. Donor-derived CML66-reactive T cells were detected at low levels in vivo in marrow before DLI by ELISpot and by a nested PCR-based assay to detect clonotypic T-cell receptor sequences but not in blood of the patient pre-DLI nor of the graft donor. CD4+ DLI results in rapid expansion of preexisting marrow-resident leukemia-specific donor CD8+ T cells, followed by a cascade of antigen-specific immune responses detectable in blood. Our single-antigen analysis thus shows that durable posttransplant tumor immunity is directed in part against nonpolymorphic overexpressed leukemia antigens that elicit coordinated cellular and humoral immunity.