MilliporeSigma
  • The IHPK1 gene is disrupted at the 3p21.31 breakpoint of t(3;9) in a family with type 2 diabetes mellitus.
The IHPK1 gene is disrupted at the 3p21.31 breakpoint of t(3;9) in a family with type 2 diabetes mellitus.
Journal of human genetics (2004-06-29)
Junichi Kamimura, Keiko Wakui, Hiroko Kadowaki, Yukio Watanabe, Kazuaki Miyake, Naoki Harada, Michiyo Sakamoto, Akira Kinoshita, Koh-ichiro Yoshiura, Tohru Ohta, Tatsuya Kishino, Mutsuo Ishikawa, Masato Kasuga, Yoshimitsu Fukushima, Norio Niikawa, Naomichi Matsumoto
ABSTRACT
Type 2 diabetes mellitus (T2DM) is a group of multifactorial disorders due to either defective insulin secretion or action. Despite the fact that numerous genetic researches of T2DM have been pursued, the pathogenic mechanisms remain obscure. We encountered a T2DM family associated with a balanced reciprocal translocation, t(3;9)(p21.31;q33.1). To isolate a candidate gene susceptible to T2DM, we constructed physical maps covering both the 3p and 9q breakpoints of the translocation in the family. Consequently, the inositol hexaphosphate kinase 1 gene ( IHPK1) (OMIM *606991) was found to be disrupted at the 3p21.31 breakpoint. We then carried out sequence analysis for all coding regions of IHPK1 in 405 unrelated T2DM patients in order to validate whether aberrations of the gene are common in T2DM patients, but we failed to detect any pathogenic changes. The disruption of IHPK1 or another predisposing gene affected by position effect of the translocation may explain the T2DM phenotype at least in this family. Alternatively, the IHPK1 disruption in the family is a chance association.