MilliporeSigma
  • An efficient nano-based theranostic system for multi-modal imaging-guided photothermal sterilization in gastrointestinal tract.
An efficient nano-based theranostic system for multi-modal imaging-guided photothermal sterilization in gastrointestinal tract.
Biomaterials (2015-05-03)
Zhen Liu, Jianhua Liu, Rui Wang, Yingda Du, Jinsong Ren, Xiaogang Qu
ABSTRACT
Since understanding the healthy status of gastrointestinal tract (GI tract) is of vital importance, clinical implementation for GI tract-related disease have attracted much more attention along with the rapid development of modern medicine. Here, a multifunctional theranostic system combining X-rays/CT/photothermal/photoacoustic mapping of GI tract and imaging-guided photothermal anti-bacterial treatment is designed and constructed. PEGylated W18O49 nanosheets (PEG-W18O49) are created via a facile solvothermal method and an in situ probe-sonication approach. In terms of excellent colloidal stability, low cytotoxicity, and neglectable hemolysis of PEG-W18O49, we demonstrate the first example of high-performance four-modal imaging of GI tract by using these nanosheets as contrast agents. More importantly, due to their intrinsic absorption of NIR light, glutaraldehyde-modified PEG-W18O49 are successfully applied as fault-free targeted photothermal agents for imaging-guided killing of bacteria on a mouse infection model. Critical to pre-clinical and clinical prospects, long-term toxicity is further investigated after oral administration of these theranostic agents. These kinds of tungsten-based nanomaterials exhibit great potential as multi-modal contrast agents for directed visualization of GI tract and anti-bacterial agents for phothothermal sterilization.
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol 5 M solution
Pricing and availability is not currently available.