MilliporeSigma
  • Osteoblast biocompatibility of novel chitosan crosslinker, hexamethylene-1,6-diaminocarboxysulfonate.
Osteoblast biocompatibility of novel chitosan crosslinker, hexamethylene-1,6-diaminocarboxysulfonate.
Journal of biomedical materials research. Part A (2015-02-18)
Laura T Beringer, Marjorie A Kiechel, Yuko Komiya, Amalie E Donius, Raymond Habas, Ulrike G K Wegst, Caroline L Schauer
ABSTRACT
Chitosan is a naturally occurring polysaccharide, which has proven to be an attractive candidate for bone tissue engineering, due to its ability to promote osteoblast mineralization. Electrospinning has become a well-established cell scaffold processing technique, as it produces a high surface area to volume fibrous material that can mimic the three dimensionality of the extracellular matrix of a cell. In this study, we have investigated the osteoblast response to two different chemically crosslinked (hexamethylene-1,6-diaminocarboxysulfonate (HDACS) and genipin) electrospun chitosan scaffolds and their film counterparts in order to determine how material chemistry influences cellular behavior in conjunction with material topology. In addition, material properties of each fiber scaffold such as porosity and tensile strength were considered. MLO-A5 osteoblast cells grown on chitosan-HDACS scaffolds were found to display a more organized cellular network, along with significantly more filopodia extensions, compared to those grown on chitosan-genipin scaffolds. After 2 days of growth on chitosan-HDACS fibers, a higher level of alkaline phosphatase expression in MLO-A5 cells was reported compared to that of either chitosan-genipin fibers or films. These results indicate that not only chemistry, but also surface topology is an important effecter of cellular behavior. Ultimately, chitosan-HDACS fiber scaffolds provided an adequate substrate for osteoblast attachment and proliferation.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%