MilliporeSigma
  • Genotoxicity evaluation of benzene, di(2-ethylhexyl) phthalate, and trisodium ethylenediamine tetraacetic acid monohydrate using a combined rat comet/micronucleus assays.
Genotoxicity evaluation of benzene, di(2-ethylhexyl) phthalate, and trisodium ethylenediamine tetraacetic acid monohydrate using a combined rat comet/micronucleus assays.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-07-28)
Sachiko Kitamoto, Ryoko Matsuyama, Yasuaki Uematsu, Keiko Ogata, Mika Ota, Toru Yamada, Kaori Miyata, Juki Kimura, Hitoshi Funabashi, Koichi Saito
ABSTRACT
As a part of the Japanese Center for the Validation of Alternative Methods (JaCVAM)-initiative international validation study of the in vivo alkaline comet assay (comet assay), we examined DNA damage in the liver, stomach, and bone marrow of rats dosed orally three times with up to 2000 mg/kg of benzene, di(2-ethylhexyl) phthalate, and trisodium ethylenediamine tetraacetic acid monohydrate. All three compounds gave negative results in the liver and stomach. In addition, a bone marrow comet and micronucleus analysis revealed that benzene, but not di(2-ethylhexyl) phthalate or trisodium ethylenediamine tetraacetic acid monohydrate induced a significant increase in the median % tail DNA and micronucleated polychromatic erythrocytes, compared with the respective concurrent vehicle control. These results were in good agreement with the previously reported genotoxicity findings for each compound. The present study has shown that combining the micronucleus test with the comet assay and carrying out these analyses simultaneously is effective in clarifying the mechanism of action of genotoxic compounds such as benzene.
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl methanesulfonate, liquid
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
SAFC
Tromethamine
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%