MilliporeSigma
  • Coleifolides A and B, Two New Sesterterpenoids from the Aerial Parts of Scutellaria coleifolia H.Lév.
Coleifolides A and B, Two New Sesterterpenoids from the Aerial Parts of Scutellaria coleifolia H.Lév.
Chemistry & biodiversity (2015-08-13)
Shin-Ichiro Kurimoto, Jian-Xin Pu, Han-Dong Sun, Yoshihisa Takaishi, Yoshiki Kashiwada
ABSTRACT
Coleifolides A and B (1 and 2), two new sesterterpenoids with a β-methyl-α,β-unsaturated-γ-lactone moiety, were isolated from the aerial parts of Scutellaria coleifolia H.Lév. (Lamiaceae), together with three known compounds. Their structures were elucidated by NMR and MS examinations. Coleifolides A and B were concluded to be partially racemic compounds by the HPLC analysis using a chiral column or introduction of chiral derivatizing agents. The absolute configuration of the major isomer was determined by analyses of the CD spectrum as well as NMR data of (R)- and (S)-2-NMA derivatives. Coleifolides A and B are structurally similar to manoalide derivatives, previously isolated from marine sponges, and appear to be the first examples of this type of compounds being isolated from higher plants.
Sigma-Aldrich
cis-4-Hydroxy-L-proline, collagen synthesis inhibitor